
Vanadium(III) chloride, Sigma-Aldrich®
: Merck
:7718-98-1
:
:Cl₃V
:MFCD00011454
:157.3 g/mol
:3 g/cm³ (20 °C)
:231-744-6
:2475
:8,
:M-Clarity™ quality level = MQ200
:GHS07
- Solubility:(20 °C) soluble
- Molar Mass:157.3 g/mol
- Bulk Density:1000 kg/m3
- Density:3.0 g/cm3 (20 °C)
- Synonyms:Vanadium trichloride
- Hill Formula:Cl3V
- Chemical Formula:VCl3
Frequently Bought Together