Tantalum pentachloride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Tantalum (V) chloride
CAS Number:7721-01-9
:
Formula:Cl₅Ta
MDL Number:MFCD00011253
Boiling Pt:242 °C
Melting Pt:216 °C
Density:3.68
Molecular Weight:358.21 g/mol
EINECS:231-755-6
UN:3260
ADR:8 III
Merck Index:14,9054
:Ampuled under argon
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together