

Titanium(IV) ethanolate for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:3087-36-3
:
Formula:C₈H₂₀O₄Ti
MDL Number:MFCD00009071
:228.12 g/mol
Boiling Pt:150 °C (1013 hPa)
Density:1,107 g/cm³ (20 °C)
Flash Pt:33 °C
Storage Temperature:Ambient
EINECS:221-410-8
ADR:3,II
:M-Clarity™ quality level = MQ200
UN:1993
:GHS02
:GHS07
Specification Test Results
- Solubility:(20 °C) Hydrolysis
- Melting Point:-40 °C
- Molar Mass:228.15 g/mol
- Boiling Point:150 - 155 °C (16 hPa)
- Flash Point:33 °C
- Refractive Index:1.5043 (20 °C, 589 nm)
- Density:1.08 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together