

Titanium(IV) butanolate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:5593-70-4
:
Formula:C₁₆H₃₆O₄Ti
MDL Number:MFCD00009433
:340.34 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:–55 °C
Density:0,993 g/cm³ (20 °C)
Flash Pt:47 °C
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:227-006-8
:GHS02
:GHS05
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:(20 °C) (slow decomposition)
- Melting Point:-40 °C
- Molar Mass:340.32 g/mol
- Boiling Point:310 - 314 °C (1013 hPa)
- Vapor Pressure:5.6 hPa (20 °C)
- Flash Point:47 °C
- Explosion Limit:2.0 - 12.0 %(V)
- Density:0.99 g/cm3 (20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.21084.0100_TDS_EN ProductPage_821084
Product specification
msds_821084
SDS
Frequently Bought Together