


Titanium(IV) butanolate 98+%
:5593-70-4
:
:C₁₆H₃₆O₄Ti
:MFCD00009433
:340.34 g/mol
:310…314 °C (760 mmHg)
:–75 °C
:0,99 g/cm³
:47 °C
:Ambient
:227-006-8
:1993
:3,III
:GHS02
:GHS05
:GHS07
- Appearance:Clear colorless to light yellow Liquid (May darken on storage)
- Infrared spectrum:Conforms
- Viscosity:50 to 90 cP (at 25°C)
- Gravimetry:23.0 to 23.9 % (TiO2)
Frequently Bought Together