Titanium(II) oxide ≥99.5% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12137-20-1
:
Formula:OTi
MDL Number:MFCD00036279
:63.88 g/mol
Boiling Pt:>3000 °C
Melting Pt:1700 °C
Density:4,95
Storage Temperature:Ambient
EINECS:235-236-5
Related Information
Frequently Bought Together