Hafnium dioxide ≥99.95% (metals basis excluding Zr) (max. 0.5% Zr)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12055-23-1
:
Formula:HfO₂
MDL Number:MFCD00003565
:210.49 g/mol
Merck Index:14,4588
Boiling Pt:5400 °C (1013 hPa)
Melting Pt:2774 °C
Density:9,68
Storage Temperature:Ambient
EINECS:235-013-2
Frequently Bought Together