

Titanium(IV) iodide, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7720-83-4
:
Formula:I₄Ti
MDL Number:MFCD00043077
Boiling Pt:360 °C (1013 hPa)
Melting Pt:150 °C
Density:4,30
Storage Temperature:Ambient
ADR:8,II
:ampouled under argon
:555.5 g/mol
EINECS:231-754-0
UN:3260
:GHS05
:GHS07
Related Information
Frequently Bought Together