Diyttrium trioxide ≥99.999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Yttrium oxide, Yttrium (III) oxide
CAS Number:1314-36-9
:
Formula:Y₂O₃
MDL Number:MFCD00011473
Boiling Pt:4300
Melting Pt:2410
Density:5.01
Molecular Weight:225.81 g/mol
Merck Index:14,10107
EINECS:215-233-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together