Order Entry
Northern Ireland
ContactUsLinkComponent
Cadmium(II) sulphate ≥99.0% ACS, Fluka™
Cadmium(II) sulphate ≥99.0% ACS, Fluka™
Catalog # 383082-500G
CAS Number:  
Cadmium(II) sulphate ≥99.0% ACS, Fluka™
Catalog # 383082-500G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Loss on drying (150 °C, 4 h):
    Max. 1.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Calcium (Ca):
    Max. 0.01 %
  • Copper (Cu):
    Max. 0.002 %
  • Iron (Fe):
    Max. 0.001 %
  • Potassium (K):
    Max. 0.02 %
  • Sodium (Na):
    Max. 0.05 %
  • Lead (Pb):
    Max. 0.003 %
  • Zinc (Zn):
    Max. 0.05 %
  • Chloride (Cl):
    Max. 0.001 %
  • Total nitrogen (as NO₃):
    Max. 0.003 %

Specification Test Results

About this item

 :
 :208.47 g/mol
Merck Index:12,01665
Melting Pt:1,000 °C
Density:4,69 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-331-6
UN:2570
ADR:6.1,III
 :GHS08
 :GHS06
 :GHS09
CAS Number:10124-36-4
Formula:CdO₄S
MDL Number:MFCD00010923