


Cadmium(II) fluoride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7790-79-6
:
Formula:CdF₂
MDL Number:MFCD00010919
:150.41 g/mol
Merck Index:14,1619
Boiling Pt:1758 °C
Melting Pt:1100 °C
Density:6,64
Storage Temperature:Ambient
EINECS:232-222-0
UN:2570
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Frequently Bought Together