

Mercury(I) chloride 99+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10112-91-1
:
Formula:Cl₂Hg₂
MDL Number:MFCD00011043
:472.09 g/mol
Merck Index:12,05951
Boiling Pt:384 °C (1013 hPa)
Melting Pt:400 °C (subl.)
Density:7,15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-307-5
UN:2025
ADR:6.1,III
:GHS06
:GHS09
Specification Test Results
- Appearance:Off-white Powder
- Titration Iodimetric:≥99.0 %
- Mercuric Chloride:≤0.01 % (HgCl2)
- Residue after reduction:≤0.02 %
Specification Test Results
Frequently Bought Together