
Silver sulphate ≥99.99% (metals basis) Ag ≥ 68.9%, Premion®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:14,8529
Boiling Pt:1085 °C (dec.)
Melting Pt:657 °C
Density:5,45
Storage Temperature:Ambient
EINECS:233-653-7
UN:3077
ADR:9,III
:GHS05
Related Information
Frequently Bought Together