

Silver sulphate 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Merck Index:13,08603
Boiling Pt:1085 °C (760 mmHg)
Melting Pt:652 °C
Density:5,45 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-653-7
UN:3077
ADR:9,III
:GHS05
:GHS09
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Titration Argentometric:≥98.5 %
- Nitrate (NO3):≤10 ppm
- Iron (Fe):≤10 ppm
- Insoluble matter:≤0.02 % (in water and AgCl)
- Subst not pptd by HCl:≤0.03 %
Specification Test Results
Frequently Bought Together