
2,7-Dihydroxynaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:582-17-2
:
Formula:C₁₀H₈O₂
MDL Number:MFCD00004085
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:185 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:209-478-7
:GHS07
Specification Test Results
- Solubility:(20 °C) practically insoluble
- Melting Point:184 - 187 °C
- Molar Mass:160.17 g/mol
- Bulk Density:600 - 700 kg/m3
- Synonyms:2,7-Dihydroxynaphthalene
- Hill Formula:C10H8O2
Specification Test Results
Frequently Bought Together