1,3-Dihydroxynaphthalene ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965
Synonyms: Naphthalene-1,3-diol, Naphthalene-1,3-diol
- CAS Number:132-86-5
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00003965
- Molecular Weight:160.17 g/mol
- Merck Index:14,6396
- Boiling Pt:386 °C (1013 hPa)
- Melting Pt:123…126 °C
- Density:1,12 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-079-7
3 Options Available Below
Product Details & Documents
Purity: 98% CAS No.: 132-86-5 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 123-126° Beilstein Reference: 2044002 Merck Reference: 14,6396 MDL No.: MFCD00003965
Synonyms: Naphthalene-1,3-diol, Naphthalene-1,3-diol
- CAS Number:132-86-5
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00003965
- Molecular Weight:160.17 g/mol
- Merck Index:14,6396
- Boiling Pt:386 °C (1013 hPa)
- Melting Pt:123…126 °C
- Density:1,12 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:205-079-7
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



