2-Methoxynaphthalene 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:93-04-9
:
Formula:C₁₁H₁₀O
MDL Number:MFCD00004061
:158.2 g/mol
Merck Index:12,06076
Boiling Pt:274 °C (1013 hPa)
Melting Pt:73…75 °C
Density:1,266 g/cm³ (20 °C)
EINECS:202-213-6
Specification Test Results
- Appearance:White to yellow-brown Crystalline solid
- Infrared spectrum:Conforms
- Melting point:≥71.5 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together