1,8-Diaminonaphthalene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:479-27-6
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004033
:158.2 g/mol
Merck Index:14,6372
Boiling Pt:205 °C (12 mmHg)
Melting Pt:61…65 °C
Density:1,18 g/cm³ (20 °C)
Flash Pt:195 °C (383 °F)
Storage Temperature:Ambient
EINECS:207-529-8
UN:3259
ADR:8,III
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together