1,8-Diaminonaphthalene 97%
:479-27-6
:
:C₁₀H₁₀N₂
:MFCD00004033
:158.2 g/mol
:12,06459
:205 °C (12 mmHg)
:63…66 °C
:1,18 g/cm³ (20 °C)
:195 °C
:Ambient
:207-529-8
:3077
:9,III
:GHS07
:GHS09
- Appearance:Grey to red to purple to dark brown Crystals or flakes and/or chunks
- Infrared spectrum:Conforms
- Melting point:61 to 66 °C
- GC:≥96.0 %
Frequently Bought Together