ContactUsLinkComponent
1-Naphthyl isothiocyanate ≥98%
1-Naphthyl isothiocyanate ≥98%
  L12418.09
 :  
1-Naphthyl isothiocyanate ≥98%
  L12418.09
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236
 :
 :185.25 g/mol
 :14,6411
 :100 °C (0,2 mmHg)
 :54…57 °C
 :1,81 g/cm³ (20 °C)
 :Ambient
 :208-990-8
 :2923
 :8,III
 :GHS06
 :GHS05
 :551-06-4
 :C₁₁H₇NS
 :MFCD00003882