Samarium(III) oxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12060-58-1
:
Formula:O₃Sm₂
MDL Number:MFCD00011237
:348.72 g/mol
Merck Index:14,8347
Melting Pt:2300 °C
Density:7,68 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:235-043-6
Related Information
Frequently Bought Together