
Digadolinium trioxide ≥99.999% (REO, rare earth oxide basis), REacton®
Catalog # 11289.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:362.5 g/mol
Merck Index:14,4326
Melting Pt:2330 °C
Density:7,407
EINECS:235-060-9
:GHS07
Storage Temperature:Ambient
CAS Number:12064-62-9
Formula:Gd₂O₃
MDL Number:MFCD00011026