Cerium(IV) oxide 1-3 mm for elementary analysis, Supelco®
1.02263.0100
: Merck
:
- Pack type:Plastic can
- Pk:100 g
- Seq:3401
- Solubility:(20 °C) insoluble
- Melting Point:2000 °C
- Molar Mass:172.11 g/mol
- Bulk Density:1280 kg/m3
- Density:7.3 g/cm3 (20 °C)
:
:172.11 g/mol
:12,02035
:3468 °C (1013 hPa)
:2000 °C
:7,13 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ100
:215-150-4
:1306-38-3
:CeO₂
:MFCD00010933