Cerium(IV) oxide 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ceric oxide, Cerium dioxide
CAS Number:1306-38-3
:
Formula:CeO₂
MDL Number:MFCD00010933
:172.11 g/mol
Merck Index:12,02035
Boiling Pt:3468 °C (1013 hPa)
Melting Pt:2600 °C
Density:7,13 g/cm³
Storage Temperature:Ambient
EINECS:215-150-4
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Powder (May give a pink to red-brown color due to traces lanthanum, neodymium, praseodymium, etc.)
- Total conc. of imp.as REO:≤0.1 %
Specification Test Results
Frequently Bought Together