Erbium triiodide, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13813-42-8
:
Formula:ErI₃
MDL Number:MFCD00016075
:547.97 g/mol
Boiling Pt:1280 °C
Melting Pt:1020 °C
Density:∼5,5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:237-471-9
Related Information
Frequently Bought Together