

Samarium(III) fluoride, anhydrous ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13765-24-7
:
Formula:F₃Sm
MDL Number:MFCD00049540
:207.36 g/mol
Boiling Pt:2323 °C (1013 hPa)
Melting Pt:1306 °C
Density:4,362 g/cm³ (2327 °C)
Storage Temperature:Ambient
EINECS:237-367-3
UN:3288
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together