Cerium trifluoride ≥99.9%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cerous fluoride, Cerium (III) fluoride
CAS Number:7758-88-5
:
Formula:CeF₃
MDL Number:MFCD00010932
Boiling Pt:∼2300 °C
Melting Pt:1460 °C
Density:6.16
Molecular Weight:197.11 g/mol
Merck Index:12,02042
EINECS:231-841-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together