
Terbium(III) chloride hexahydrate ≥99.999% (metals basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13798-24-8
:
Formula:Cl₃Tb·6H₂O
MDL Number:MFCD00149870
:373.38 g/mol
Merck Index:14,9157
Melting Pt:145 °C
Density:4,35 g/cm³ (20 °C)
EINECS:233-132-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together