
Samarium trichloride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10361-82-7
:
Formula:Cl₃Sm
MDL Number:MFCD00011235
:256.72 g/mol
Merck Index:14,8347
Boiling Pt: Dec.
Melting Pt:678 °C
Density:4,46
EINECS:233-797-0
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together