
Neodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10024-93-8
:
Formula:Cl₃Nd
MDL Number:MFCD00011132
:250.6 g/mol
Merck Index:14,6450
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:784 °C
Density:4,134
:ampouled under argon
EINECS:233-031-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together