
Neodymium trichloride, anhydrous 99.9% (trace metals basis)
Catalog # ACRO315070050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light green Powder
- Total conc. of imp.as REO:≤0.1 %
- Neodymium (Nd):55.8 to 59.3 %
Specification Test Results
About this item
:
:250.6 g/mol
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:784 °C
Density:4,13 g/cm³ (20 °C)
EINECS:233-031-5
:GHS07
CAS Number:10024-93-8
Formula:Cl₃Nd
MDL Number:MFCD00011132