
Erbium trichloride, anhydrous ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10138-41-7
:
Formula:Cl₃Er
MDL Number:MFCD00010989
:273.62 g/mol
Boiling Pt:1500 °C
Melting Pt:774 °C
Density:4,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-385-0
:GHS07
Related Information
Frequently Bought Together