Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methylindole
6-Methylindole
6-Methylindole
MOLE43956247-250MG
:
Molekula
:
6-Methylindole
MOLE43956247-250MG
:
Molekula
:
Pk:
250 mg
Seq:
2590
:
:
131.18 g/mol
:
260 °C (1013 hPa)
:
30…31 °C
:
1,05 g/cm³ (20 °C)
:
3334
:
9,
:
GHS07
:
3420-02-8
:
C₉H₉N
:
MFCD00005682