

Indole for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:120-72-9
:
Formula:C₈H₇N
MDL Number:MFCD00005607
:117.15 g/mol
Merck Index:13,04985
Boiling Pt:254…255 °C (1013 hPa)
Melting Pt:50…51 °C
Density:1,22 g/cm³ (20 °C)
EINECS:204-420-7
UN:2811
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:110 °C
:GHS06
:GHS09
Specification Test Results
- Solubility:3.560 g/l (25 °C)
- Melting Point:52 °C
- Molar Mass:117.15 g/mol
- Bulk Density:230 kg/m3
- Boiling Point:254 °C
- Vapor Pressure:0.016 hPa (25 °C)
- Flash Point:110 °C
- Density:1.22 g/cm3 (20 °C)
- pH:5.9 (1000 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Frequently Bought Together