£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
2-Bromo-1-indanone
2-Bromo-1-indanone tech. 85%, Technical Grade
2-Bromo-1-indanone tech. 85%, Technical Grade
L12453.06
:
Thermo Fisher Scientific
:
2-Bromo-1-indanone tech. 85%, Technical Grade
L12453.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
211.06 g/mol
:
110…112 °C (0,4 mmHg)
:
37…39 °C
:
GHS07
:
Ambient
:
1775-27-5
:
C₉H₇BrO
:
MFCD00041030