
1,1'-Thiocarbonyldiimidazole tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6160-65-2
:
Formula:C₇H₆N₄S
MDL Number:MFCD00005289
:178.22 g/mol
Melting Pt:98…102 °C
EINECS:228-183-4
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together