1,2-Dimethylimidazole 96+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: 1,2-Dimethyl-1H-imidazole
- CAS Number:1739-84-0
- Formula:C₅H₈N₂
- MDL Number:MFCD00005294
- Molecular Weight:96.13 g/mol
- Boiling Pt:204 °C (1013 hPa)
- Melting Pt:37…39 °C
- Density:1,084 g/cm³
- Flash Pt:93 °C
- Storage Temperature:Ambient
- EINECS:217-101-2
2 Options Available Below
Product Details & Documents
Synonyms: 1,2-Dimethyl-1H-imidazole
- CAS Number:1739-84-0
- Formula:C₅H₈N₂
- MDL Number:MFCD00005294
- Molecular Weight:96.13 g/mol
- Boiling Pt:204 °C (1013 hPa)
- Melting Pt:37…39 °C
- Density:1,084 g/cm³
- Flash Pt:93 °C
- Storage Temperature:Ambient
- EINECS:217-101-2
Documents
ACRO15944
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White to yellow to orange Crystals or crystalline mass or liquid (May darken on storage)
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Insoluble matter:≤0.05 % (In methanol)
- Residue after reduction:≤0.02 %



