Imidazole
Supplier: Molekula
Certificates
About this item
Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Boiling Pt:255
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Molecular Weight:68.08 g/mol
Flash Pt:135
Storage Temperature:Ambient
UN:2923
ADR:8 III
Merck Index:13,04935
EINECS:206-019-2
:GHS05
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together