
2-Nitroimidazole
Catalog # MOLE13834750-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:113.08 g/mol
Merck Index:13,00923
Melting Pt:279…284 °C
Storage Temperature:Ambient
EINECS:208-425-5
UN:2811
ADR:6.1,III
:GHS07
CAS Number:527-73-1
Formula:C₃H₃N₃O₂
MDL Number:MFCD00005185