

Dimethyl sulphide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-18-3
:
Formula:C₂H₆S
MDL Number:MFCD00008562
:62.14 g/mol
Merck Index:12,06204
Boiling Pt:36,2 °C (1013 hPa)
Melting Pt:–98 °C
Density:0,85 g/cm³ (20 °C)
EINECS:200-846-2
UN:1164
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:–18 °C
:GHS02
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:2 g/l (20 °C)
- Melting Point:-98 °C
- Molar Mass:62.14 g/mol
- Boiling Point:37 °C (1013 hPa)
- Vapor Pressure:530 hPa (25 °C)
- Flash Point:-48 °C
- Refractive Index:1.4351 (20 °C, 589 nm)
- Explosion Limit:2.2 - 19.7 %(V)
- Density:0.85 g/cm3 (20 °C)
- Ignition Point:205 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
Frequently Bought Together