


Selenium disulphide ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7488-56-4
:
Formula:S₂Se
MDL Number:MFCD00011227
:143.09 g/mol
Merck Index:14,8436
Boiling Pt: Dec.
Melting Pt:<100 °C
Density:0,03 g/cm³ (–65 °C)
Storage Temperature:Ambient
EINECS:231-303-8
UN:2657
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Related Information
Frequently Bought Together