Order Entry
Northern Ireland
ContactUsLinkComponent
Ammonium thiocyanate
Ammonium thiocyanate
Catalog # MOLE18466896-1KG
Supplier:  Molekula
CAS Number:  
Ammonium thiocyanate
Catalog # MOLE18466896-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :76.12 g/mol
Merck Index:13,00564
Melting Pt:150 °C
Density:1,305 g/cm³ (20 °C)
EINECS:217-175-6
 :GHS07
Storage Temperature:Ambient
CAS Number:1762-95-4
Formula:NH₄SCN
MDL Number:MFCD00011428