


Ammonium peroxodisulphate (APS) ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1,98 g/cm³ (20 °C)
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS03
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 98.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue:Max. 0.05 %
- Insolubility in water:Max. 0.02 %
- Cl (Chloride):Max. 5 ppm
- Fe (Iron):Max. 10 ppm
- Mn (Manganese):Max. 2 ppm
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC21300.260 spec_VWRC21300.293
Product specification
Frequently Bought Together