Ammonium peroxodisulphate (APS), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1,98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-786-5
ADR:5.1,III
:M-Clarity™ quality level = MQ200
UN:1444
:GHS03
:GHS08
:GHS07
Frequently Bought Together