


Ammonium peroxodisulphate (APS) ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:14,541
Melting Pt:120 °C (dec.)
Density:1,982
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together