Ammonium peroxodisulphate (APS) 98%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-54-0
:
Formula:(NH₄)₂S₂O₈
MDL Number:MFCD00003390
:228.2 g/mol
Merck Index:13,00545
Melting Pt:120 °C
Density:1,98 g/cm³
EINECS:231-786-5
UN:1444
ADR:5.1,III
:GHS07
:GHS08
:GHS03
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Titration with KMnO4:≥97.5 %
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤50 ppm
Specification Test Results
Frequently Bought Together