
Ammonium carbonate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0015.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (NH3):min. 30.0 %
- Trace Impurities (in ppm)Chloride (Cl):max. 5
- Trace Impurities (in ppm)Iron (Fe):max. 5
- Trace Impurities (in ppm)Heavy Metals (as Pb):max. 5
- Insoluble Matter:max. 0.005 %
- Non-volatile Matter:max. 0.01 %
- Sulfur Compounds (as SO4):max. 0.002 %
Specification Test Results
About this item
:
:96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1,5 g/cm³ (20 °C)
EINECS:208-058-0
:GHS07
Storage Temperature:Ambient
CAS Number:506-87-6
:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890