
Ammonium bromide 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12124-97-9
:
Formula:H₄BrN
MDL Number:MFCD00011419
:97.94 g/mol
Merck Index:13,00505
Boiling Pt:235 °C (760 mmHg)
Melting Pt:452 °C (subl.)
Density:2,42 g/cm³ (20 °C)
EINECS:235-183-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Titration Argentometric:≥99.0 % (on dry substance)
- Loss on drying:≤0.2 % (105°C)
- Sulfated ash:≤0.05 %
- pH:4.5 to 6.0 (5 % solution)
- Chloride (Cl):≤0.2 %
- Bromate (BrO3):≤0.001 %
- Sulfate (SO4):≤0.005 %
- Barium (Ba):≤20 ppm
- Copper (Cu):≤2 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤2 ppm
- Nickel (Ni):≤2 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
Frequently Bought Together