
Tetraphenylphosphonium bromide 99%
:2751-90-8
:
:C₂₄H₂₀BrP
:MFCD00011915
:295…299 °C
:260 °C
:419.3 g/mol
:220-393-4
:GHS07
:Ambient
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:296 to 300 °C
- Titration Argentometric:≥98.5 %
Frequently Bought Together