Order Entry
Northern Ireland
ContactUsLinkComponent
 
 
Tris(2-methylphenyl)phosphine for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Tris(2-methylphenyl)phosphine for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Melting Point:
    124 - 127 °C
  • Molar Mass:
    304.38 g/mol
  • Boiling Point:
    238 °C (15 hPa)

 

Frequently Bought Together